Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Original Fit Work Pants, Dark Navy, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

874DN32UU 874DN4031 874DN5230 874DN3631 874DN34UU 874DN4634 874DN3228 874DN4432 874DN4630 874DN3034 874DN50UU 874DN5032 874DN5030 874DN3432 874DN5630 874DN3828 874DN3029 874DN4830 874DN4429 874DN5830 874DN58UU 874DN72UU 874DN2832 874DN3836 874DN36UU 874DN4231 874DN64UU 874DN4834 874DN3832 874DN68UU 874DN3331 874DN4832 874DN3436 874DN44UU 874DN3830 874DN66UU 874DN4030 874DN3428 874DN3429 874DN42UU 874DN44UL 874DN3829 874DN3831 874DN3134 874DN5832 874DN40UU 874DN3232 874DN3634 874DN4034 874DN34UL 874DN3234 874DN5432 874DN30UL 874DN3834 874DN5430 874DN3632 874DN3628 874DN2932 874DN3434 874DN4434 874DN30UU 874DN4029 874DN70UU 874DN28UU 874DN3334 874DN4028 874DN3030 874DN3230 874DN3636 874DN32UL 874DN4032 874DN5034 874DN4229 874DN3332 874DN40UL 874DN38UU 874DN3129 874DN2930 874DN3229 874DN3132 874DN46UU 874DN3330 874DN4431 874DN3630 874DN60UU 874DN3430 874DN3031 874DN2929 874DN3032 874DN56UU 874DN62UU 874DN2830 874DN4234 874DN28UL 874DN36UL 874DN3629 874DN52UU 874DN5232 874DN4428 874DN54UU 874DN3130 874DN4430 874DN3131 874DN5632 874DN4232 874DN3231 874DN4632 874DN3431 874DN38UL 874DN48UU 874DN4228 874DN42UL 874DN3329 874DN4230
76148-128EA 78.84 USD InStock
76148-128 76148-134 76150-508 76148-098 76148-132 76150-506 76148-138 76150-504 76148-136 76148-092 76150-502 76150-468 76148-090 76150-500 76150-466 76148-096 76148-130 76150-464 76148-094 76150-570 76148-118 76148-122 76148-120 76148-088 76148-126 76148-124 76150-578 76148-082 76148-080 76150-576 76150-574 76148-086 76150-572 76148-084 76150-560 76150-568 76150-566 76150-564 76150-562 76148-144 76148-142 76148-146 76150-558 76150-556 76150-554 76148-140 76150-552 76150-550 76148-056 76148-054 76150-548 76148-058 76150-546 76150-544 76148-052 76150-542 76150-540 76148-050 76150-494 76150-492 76150-490 76148-046 76148-044 76150-538 76150-536 76148-048 76150-534 76150-532 76150-498 76150-530 76148-042 76150-496 76148-040 76150-482 76150-480 76148-108 76148-106 76148-078 76148-112 76148-076 76148-110 76150-528 76148-116 76150-526 76148-114 76148-070 76150-524 76150-522 76150-488 76150-520 76148-074 76150-486 76148-072 76150-484 76150-472 76150-470 76148-100 76148-068 76148-066 76150-518 76148-104 76150-516 76148-102 76150-878 76150-514 76150-876 76150-512 76148-060 76150-478 76150-510 76150-476 76148-064 76150-474 76148-062
Original Fit Work Pants, Dark Navy, Dickies®
Workwear
Dickies® style 874.

  • Wrinkle resistant
  • Stain resistant
  • Center crease
  • Tunnel belt loops
  • Made of 8.5 oz. twill, 65% polyester/35% cotton

Sizing Chart: Size measurements indicate the waist by inseam, in inches. UU size designations represent a 37" inseam, unhemmed. UL size designations represent a 39" inseam, unhemmed.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty