Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Original Fit Work Pants, Lincoln Green, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

874LN4230 874LN3830 874LN4032 874LN4430 874LN4632 874LN4029 874LN4030 874LN3632 874LN3432 874LN3132 874LN3428 874LN4034 874LN3030 874LN3234 874LN4232 874LN5030 874LN3032 874LN3230 874LN4630 874LN3630 874LN3332 874LN3134 874LN3232 874LN3330 874LN4830 874LN4432 874LN3430 874LN3629 874LN3130 874LN3434 874LN3834 874LN3634 874LN3034 874LN4832 874LN3334 874LN3829 874LN3832
76148-428EA 72.57 USD InStock
76148-428 76148-426 76148-448 76148-452 76148-474 76148-430 76148-472 76148-450 76148-434 76148-478 76148-456 76148-476 76148-432 76148-454 76148-470 76148-416 76148-438 76148-458 76148-436 76148-418 76148-464 76148-442 76148-420 76148-486 76148-440 76148-484 76148-462 76148-468 76148-424 76148-446 76148-466 76148-444 76148-488 76148-422 76148-482 76148-460 76148-480
Original Fit Work Pants, Lincoln Green, Dickies®
Workwear
Dickies® style 874.

  • Wrinkle resistant
  • Stain resistant
  • Center crease
  • Tunnel belt loops

Made of 8.5 oz. twill, 65% polyester/35% cotton.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty