Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Original Fit Work Pants, Olive Green, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

874OG4232 874OG2932 874OG3430 874OG4830 874OG4832 874OG3832 874OG3428 874OG3230 874OG4230 874OG3330 874OG3632 874OG3032 874OG2832 874OG3334 874OG4032 874OG4030 874OG4430 874OG4034 874OG3630 874OG3134 874OG3332 874OG3634 874OG3234 874OG3034 874OG3132 874OG4630 874OG4432 874OG3030 874OG3434 874OG3830 874OG3432 874OG3232 874OG3130 874OG4632 874OG5030 874OG3834
76148-708EA 72.57 USD InStock
76148-708 76148-746 76148-724 76148-702 76148-744 76148-700 76148-722 76148-728 76148-706 76148-704 76148-726 76148-748 76148-750 76148-694 76148-692 76148-754 76148-732 76148-710 76148-698 76148-730 76148-752 76148-696 76148-690 76148-714 76148-736 76148-758 76148-756 76148-712 76148-734 76148-718 76148-716 76148-738 76148-688 76148-742 76148-720 76148-740
Original Fit Work Pants, Olive Green, Dickies®
Workwear
Dickies® style 874.

  • Wrinkle and stain resistant
  • Center crease
  • Tunnel belt loops

Made of 8.5 oz. twill, 65% polyester/35% cotton.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty