Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Unlined Eisenhower Jackets, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

JT75DNXLRG JT75DNMRG JT75CHXLRG JT75BK3XRG JT75DN5XRG JT75CHLRG JT75DNSRG JT75DN3XRG JT75CHMRG JT75DN2XRG JT75BKMRG JT75BK2XRG JT75BK5XRG JT75DN6XRG JT75CHSRG JT75DNLRG JT75BKXLRG JT75BKLRG JT75DN4XRG JT75BKSRG JT75CH2XRG JT75BK4XRG JT75BK6XRG
76149-472EA 110.38 USD InStock
76149-472 76149-450 76149-482 76149-460 76149-474 76149-452 76149-484 76149-462 76149-454 76149-476 76149-464 76149-486 76149-478 76149-456 76149-488 76149-466 76149-444 76149-470 76149-480 76149-458 76149-446 76149-468 76149-448
Unlined Eisenhower Jackets, Dickies®
Workwear
Dickies® style JT75.

  • Slash front pockets
  • Pencil pocket on left sleeve
  • Heavy-duty brass zipper front closure
  • Adjustable tabs at waistband
  • Felled seams
  • Made of 8 oz. vat dyed twill, 65% polyester/35% cotton

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty