Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Insulated Team Jackets, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

JTC2DN3XRG JTC2BKXLRG JTC2BKMRG JTC2DNMRG JTC2DN2XRG JTC2DNLRG JTC2BK3XRG JTC2DNXLRG JTC2DN4XRG JTC2BK2XRG JTC2BK4XRG JTC2BKLRG
76149-494EA 151.4 USD InStock
76149-494 76149-496 76149-498 76149-500 76149-510 76149-502 76149-512 76149-504 76149-506 76149-490 76149-508 76149-492
Insulated Team Jackets, Dickies®
Workwear
Dickies® style JTC2.

  • Pencil pocket on the sleeve
  • Knit cuffs and collar
  • Holds color through repeated washings
  • Shell: 8 oz. vat dyed twill, 65% polyester/35% cotton
  • Lining: 4 oz. nylon taffeta quilted to polyurethane foam
  • Slash front pockets

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty