Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sanded Duck Insulated Bib Overalls, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

TB244RBDXLRG TB244RBD5XRG TB244RBKMTL TB244RBDLRG TB244RBD3XTL TB244RBDXLTL TB244RBKXLS TB244RBDMS TB244RBDLS TB244RBK2XTL TB244RBD2XTL TB244RBK3XRG TB244RBD2XS TB244RBK2XS TB244RBK3XS TB244RBKLTL TB244RBD3XS TB244RBDXLS TB244RBKLS TB244RBKXLRG TB244RBKSRG TB244RBK2XRG TB244RBD3XRG TB244RBKLRG TB244RBK5XRG TB244RBD2XRG TB244RBDSRG TB244RBKMRG TB244RBKXLTL TB244RBD4XRG TB244RBDMTL TB244RBK4XRG TB244RBKMS TB244RBDLTL TB244RBK3XTL
76149-550EA 198.72 USD InStock
76149-550 76149-530 76149-552 76149-532 76149-554 76148-996 76150-768 76149-524 76150-766 76149-546 76150-764 76149-548 76149-526 76149-528 76150-762 76150-760 76149-540 76149-542 76149-520 76149-522 76149-544 76148-998 76150-758 76150-756 76150-754 76150-776 76149-534 76150-774 76149-536 76149-514 76149-538 76149-516 76150-772 76149-518 76150-770
Sanded Duck Insulated Bib Overalls, Dickies®
Workwear
Dickies® style TB244 has double knees with clean out opening.

  • Light and strong fabric for ease of movement and comfort
  • Zip to knee
  • High cross back style
  • Elastic front straps
  • Triple needle felled seams
  • 8.5 oz. sanded duck; 100% cotton

Chest lining: 3.25 oz. 100% polyester brushed tricot; leg lining: 2 oz. 100% polyester taffeta quilted to 3.35 oz. polyester fill.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty