Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sanded Duck Insulated Coveralls, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

TV243RBKXLRG TV243RBDLTL TV243RBKMRG TV243RBDLS TV243RBKMS TV243RBK3XRG TV243RBD3XTL TV243RBDMS TV243RBK2XRG TV243RBDSRG TV243RBK2XS TV243RBD2XTL TV243RBK2XTL TV243RBDXLS TV243RBDXLTL TV243RBD3XRG TV243RBK3XTL TV243RBKXLS TV243RBD2XS TV243RBDXLRG TV243RBDMRG TV243RBDLRG TV243RBDMTL TV243RBKLS TV243RBD2XRG TV243RBKSRG TV243RBKLTL TV243RBKXLTL TV243RBKLRG TV243RBKMTL
76149-890EA 242.89 USD InStock
76149-890 76149-892 76149-894 76149-896 76149-906 76149-928 76149-908 76150-848 76150-846 76150-844 76149-920 76150-842 76149-900 76149-922 76150-840 76149-888 76149-902 76149-924 76149-926 76149-904 76149-916 76149-918 76150-854 76149-898 76149-930 76150-852 76149-910 76150-850 76149-912 76149-914
Sanded Duck Insulated Coveralls, Dickies®
Workwear
Dickies® style TV243.

  • Zip to knee, double knees with clean out opening
  • Knit storm cuffs and corduroy collar
  • Triple needle felled seams
  • Convenient pass through openings
  • Heavy-duty two-way center front zipper
  • 8.5 oz. sanded duck, 100% cotton
  • Lining: 3.25 100% polyester tricot quilted to 4 oz. polyester fill

Light and strong fabric are used for ease of movement and comfort.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty