Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thermal Lined Fleece Hoodies, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

TW382DN5X TW382DNL TW382AGXT TW382DN3T TW382BK3X TW382DBXL TW382DB3X TW382DBS TW382BKLT TW382DB5X TW382DBM TW382DN3X TW382BKL TW382AGL TW382AG5X TW382BK4X TW382AG2X TW382DB4X TW382BK3T TW382BKXT TW382BK5X TW382DNM TW382DN2T TW382AG4X TW382DNLT TW382AG3T TW382DBLT TW382DNXL TW382BK2T TW382AGLT TW382BKM TW382AGXL TW382BK2X TW382DB2X TW382DB3T TW382BKS TW382AGM TW382AG2T TW382DB2T TW382AGS TW382DNXT TW382DNS TW382DBL TW382AG3X TW382BKXL TW382DN2X TW382DBXT TW382DN4X
76149-990EA 126.19 USD InStock
76149-990 76149-992 76149-970 76149-972 76149-950 76149-994 76151-366 76151-364 76150-008 76149-942 76149-964 76149-986 76150-006 76149-988 76149-944 76149-966 76150-004 76149-968 76149-946 76150-002 76149-948 76150-000 76150-010 76149-980 76149-960 76149-982 76149-962 76149-940 76149-984 76149-938 76150-858 76150-856 76150-018 76149-996 76149-952 76149-974 76150-016 76149-954 76149-932 76149-976 76149-998 76150-014 76149-934 76149-978 76149-956 76150-012 76149-936 76149-958
Thermal Lined Fleece Hoodies, Dickies®
Workwear
Dickies® style TW382.

  • Full front zipper
  • Two deep hand-warmer pockets
  • Drawstring hood
  • Extended sizing includes big and tall
  • 7.5 oz./260 g fleece, 80% cotton/20% polyester
  • Lining: 5.8 oz./210 g thermal knit 60% cotton/40% polyester

A big favorite with repeat customers. This durable hoodie is fully lined (including hood) with waffle knit thermal fabric that traps warm air between yarns, making it warm without being too heavy. Built to last from soft fleece with hand-warmer pockets, full metal zipper, and drawstring hood.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty