Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Midweight Fleece Zip Hoodies, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

TW391DHXLTL TW391BKLTL TW391DN2XTL TW391DH3XTL TW391DH3XRG TW391DHLTL TW391BK2XRG TW391BK4XRG TW391DHXLRG TW391DHLRG TW391HGLTL TW391BKXLTL TW391DHSRG TW391DN4XTL TW391BK3XRG TW391BKSRG TW391HG4XTL TW391BK3XTL TW391HG3XTL TW391DNLTL TW391DNXLTL TW391BKLRG TW391BK4XTL TW391DH4XTL TW391BKMRG TW391BKXLRG TW391DH5XRG TW391DHMRG TW391DH2XRG TW391DH2XTL TW391DH4XRG TW391HG2XTL TW391DN3XTL TW391BK5XRG TW391BK2XTL TW391HGXLTL
76151-408EA 104.11 USD InStock
76151-408 76150-120 76150-086 76150-064 76150-042 76150-062 76150-040 76150-082 76150-060 76150-080 76150-108 76150-126 76150-048 76150-046 76150-068 76150-100 76150-066 76150-044 76150-054 76150-076 76150-074 76150-052 76150-072 76150-094 76150-050 76151-414 76150-070 76150-090 76151-410 76151-412 76150-116 76150-058 76150-112 76150-134 76150-056 76150-078
Midweight Fleece Zip Hoodies, Dickies®
Workwear
Dickies® style TW391.

  • Midweight 10.5 oz./350 g
  • Dark heather gray fabric: 55% cotton/45% polyester
  • Black fabric: 50% cotton/50% polyester
  • Two deep hand-warmer pockets
  • Drawstring 3-piece head-hugging hood for best fit
  • Extended sizing includes big and tall

This may be the most versatile piece of outerwear you'll ever own. It's warm, without being bulky, and layers well. Soft fleece made of easy-care cotton/polyester with heavy-duty full front zipper, rib knit cuffs and waistband.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty