Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Long Sleeve Heavyweight Crew Neck T-Shirts, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

WL450WHM WL450IHXT WL450CBLT WL450DSXL WL450ERXT WL450MSLT WL450IH3X WL450GH2T WL450AG2T WL450CB3T WL450CBM WL450WHXL WL450IH3T WL450KEL WL450OR2T WL450BK3T WL450AG3X WL450BKM WL450DNXL WL450CHXL WL450IHXL WL450DSXT WL450MSXT WL450DN2X WL450DS2X WL450ER3T WL450HG3T WL450WH2X WL450GH2X WL450BYM WL450WHLT WL450IHM WL450ORL WL450KE2X WL450HGL WL450CH2X WL450AG3T WL450HGLT WL450GHXT WL450CHM WL450CB3X WL450IHL WL450ORXL WL450IHLT WL450KEXT WL450DNXT WL450ORXT WL450MSL WL450WH3T WL450HGXL WL450MS3X WL450WH2T WL450HGXT WL450BYLT WL450WH3X WL450IH2X WL450BY3T WL450BK2T WL450GHL WL450ER3X WL450BKXL WL450MS2X WL450WHXT WL450DSM WL450CBXT WL450KE2T WL450GH3T WL450AG2X WL450AGXL WL450ERLT WL450CH2T WL450ORLT WL450AGXT WL450AGLT WL450MS2T WL450HG2X WL450CBXL WL450DSL WL450ERM WL450BY3X WL450BKXT WL450DS3T WL450AGM WL450DNL WL450GHLT WL450DNM WL450HG2T WL450KE3T WL450BY2T WL450CHXT WL450ERXL WL450MS3T WL450DN3T WL450AGL WL450GHM WL450DS3X WL450KE3X WL450BK3X WL450DNLT WL450CHL WL450ERL WL450CB2X WL450IH2T WL450BYXT WL450KEM WL450CH3X WL450GHXL WL450OR3X WL450CBL WL450ER2X WL450GH3X WL450BKL WL450KEXL WL450DSLT WL450CHLT WL450MSM WL450DN3X WL450CH3T WL450OR3T WL450DS2T WL450OR2X WL450MSXL WL450BYXL WL450HG3X WL450WHL WL450ORM WL450DN2T WL450HGM WL450BKLT WL450BY2X WL450BYL WL450KELT WL450CB2T WL450BK2X WL450ER2T
76150-340EA 41.01 USD InStock
76150-340 76151-426 76151-428 76151-422 76151-424 76151-420 76150-868 76150-866 76150-228 76150-348 76150-864 76150-226 76150-346 76150-224 76150-862 76150-344 76150-860 76150-342 76150-296 76151-418 76150-294 76150-292 76150-290 76151-416 76150-338 76150-612 76150-336 76150-334 76150-332 76150-298 76150-330 76150-284 76150-282 76150-280 76150-328 76150-326 76150-324 76150-322 76150-288 76150-320 76150-286 76150-274 76150-394 76150-272 76150-392 76150-270 76150-390 76150-318 76150-316 76150-314 76150-312 76150-278 76150-310 76150-398 76150-276 76150-430 76150-396 76150-384 76150-262 76150-382 76150-260 76150-380 76150-308 76150-428 76150-306 76150-426 76150-304 76150-424 76150-302 76150-422 76150-268 76150-388 76150-300 76150-266 76150-420 76150-386 76150-264 76150-252 76150-372 76150-250 76150-370 76150-418 76150-416 76150-414 76150-258 76150-412 76150-378 76150-256 76150-410 76150-376 76150-254 76150-374 76150-362 76150-240 76150-360 76151-444 76151-446 76151-440 76151-442 76151-282 76150-408 76150-406 76151-280 76150-404 76150-402 76150-248 76150-368 76150-246 76150-400 76150-366 76150-244 76150-364 76150-242 76150-230 76150-350 76151-436 76151-438 76151-278 76151-432 76151-434 76151-274 76151-430 76151-276 76151-272 76150-238 76150-358 76150-236 76150-874 76150-356 76150-234 76150-872 76150-354 76150-870 76150-232 76150-352
Long Sleeve Heavyweight Crew Neck T-Shirts, Dickies®
Workwear
Dickies® style WL450.

  • Heavyweight pocket tee
  • Chest pocket
  • Tagless back neck label for comfort
  • Taped neck and shoulder seams for longer wear
  • Ash gray: 99% cotton/1% polyester
  • Heather gray: 90% cotton/10% polyester
  • All other colors: 6.75 oz./220 g; 100% cotton

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty