Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Insulated Eisenhower Jackets, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

TJ15CHLRG TJ15KH3XRG TJ15DB5XRG TJ15DN4XRG TJ15KHXLRG TJ15DBXLLN TJ15CHMLN TJ15DNMRG TJ15BKXLLN TJ15BKMRG TJ15KHXLLN TJ15DNXLLN TJ15DB3XRG TJ15DB4XRG TJ15CHXLRG TJ15KH4XRG TJ15CH4XRG TJ15DN2XRG TJ15KHLRG TJ15DBLRG TJ15BK3XLN TJ15DNMLN TJ15BK4XRG TJ15DB3XLN TJ15KHMRG TJ15DNXLRG TJ15BKLLN TJ15DNSRG TJ15BK3XRG TJ15DNLLN TJ15KH2XLN TJ15KHMLN TJ15BKXLRG TJ15DBLLN TJ15BK2XRG TJ15CHXLLN TJ15DBSRG TJ15DBXLRG TJ15DN3XRG TJ15KHLLN TJ15DN3XLN TJ15CH2XLN TJ15CH3XRG TJ15DB2XLN TJ15DBMRG TJ15CHLLN TJ15KHSRG TJ15BKLRG TJ15BKMLN TJ15DNLRG TJ15DB2XRG TJ15DN2XLN TJ15CHMRG TJ15DN5XRG TJ15BK5XRG TJ15BK2XLN TJ15CH3XLN TJ15BKSLN TJ15KH2XRG TJ15KH3XLN TJ15DBMLN TJ15CHSRG TJ15CH2XRG TJ15BKSRG TJ15DN6XRG
76149-692EA 126.19 USD InStock
76149-692 76149-694 76149-650 76149-696 76149-652 76149-698 76149-654 76149-730 76149-690 76150-828 76150-826 76150-824 76149-722 76149-644 76149-688 76149-724 76149-646 76149-726 76149-648 76149-728 76149-680 76149-682 76149-684 76149-640 76149-720 76149-642 76149-686 76149-718 76149-678 76149-710 76149-712 76149-714 76149-716 76149-670 76149-672 76149-674 76149-750 76149-676 76149-752 76149-708 76149-700 76149-744 76149-666 76149-702 76149-746 76149-668 76149-704 76149-748 76149-706 76149-780 76149-782 76149-660 76149-784 76149-740 76149-662 76149-742 76149-786 76149-664 76149-656 76149-732 76149-658 76150-830 76149-734 76149-736 76149-738
Insulated Eisenhower Jackets, Dickies®
Workwear
Dickies® style TJ15.

  • Lightweight but warm
  • Adjustable tabs at waistband
  • Brass zipper front closure
  • Two-piece sleeve
  • Shell: 8 oz. vat dyed twill, 65% polyester/35% cotton
  • Lining: 4 oz. nylon taffeta lining quilted to polyurethane foam

These most popular Dickies® jackets are features with pencil pocket on left sleeve and slash front welt pockets. Inspired by the tough, quietly authoritative jacket favored by the revered general and president Eisenhower, this jacket is updated for today's toughest work situations. Known for its classic look, rugged durability, and comfort, it never goes out of style and is guaranteed to work as hard.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty