Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sherpa Lined Fleece Hoodies, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

TW357BKL TW357KEL TW357DH2X TW357YG2X TW357KE2X TW357BKM TW357BKS TW357YGM TW357BK2X TW357BK3X TW357BK2T TW357DHS TW357BKXL TW357KE3X TW357BKXT TW357DHL TW357DH3T TW357DHM TW357DHXT TW357KEXL TW357YGL TW357DHLT TW357DH2T TW357YG3X TW357DH3X TW357KEM TW357YGXL TW357DHXL TW357BK3T TW357BKLT
76150-020EA 160.85 USD InStock
76150-020 76151-404 76151-406 76151-400 76151-388 76151-402 76151-368 76151-384 76151-386 76151-380 76151-382 76150-028 76150-026 76150-024 76150-022 76150-032 76150-030 76151-378 76151-396 76151-374 76151-398 76151-376 76151-392 76151-370 76151-372 76151-394 76151-390 76150-038 76150-036 76150-034
Sherpa Lined Fleece Hoodies, Dickies®
Workwear
Dickies® style TW357.

  • Center front zipper closure
  • Rib knit waistband and cuff
  • Three-piece lined hood
  • 350 g, 55% cotton/45% polyester (Dark heather is 70% cotton/30% polyester)
  • Sherpa lining: 100% polyester
  • Sleeve lining: Polyester taffeta with polyester fill

Three-piece lined hood and Center front zipper closure.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty