Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, May 25. 
Whirl-Pak® Polyurethane Surface Sampling Bags with PolySponge™, Nasco
Supplier: Nasco
Whirl-Pak®
B01590
76151-838BX 229.14 USD InStock
76151-838
Whirl-Pak® Polyurethane Surface Sampling Bags with PolySponge™, Nasco
Bags Sample Bags
Make environmental surface sampling faster and easier with Whirl-Pak polyurethane surface sampling bags with premoistened sponge. The durable high-density polyurethane sponge is ready to use and resists tearing and fraying.
  • Each sponge is packed in a sterile, sealed write-on bag
  • Write-on label makes identifying samples easy and convenient
  • Sponge is hydrated with 10 ml of HiCap™ neutralizing broth
  • Terminally gamma processed to guarantee sterility (+1 Sterilization)
  • Opening Tyvek® Pull Tabs are perpendicular to the opening of the bag: Keeps your hands away for a safe, aseptic collection technique each time
To use the sponge, tear open the top of the Whirl-Pak® bag, squeeze out any excess HiCap™ Neutralizing Broth, swab the surface, insert the sponge back into the bag, fold the top of the bag over three or more times, and bend in the wire tabs to close. Send to a lab for testing.
Shelf life from sterilisation date found on the sterility documentation: 2 years from sterilised date found on your sterility confirmation due to the buffer. 2 year shelf-life with ambient storage at 2 to 30 °C. The bags are packed in a foil liner bag to prevent buffer evaporation.
Certifications: Approved by USDA, FDA, EPA and HACCP. Food safe.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty