Would you like to log in again?
VWR® Turbidity Meter
Supplier: Avantor
VWR®
76152-030
76152-030KT 2258.05 USD InStock
76152-030
VWR® Turbidity Meter
Turbidity Meters
Autoranging from 0 to 1000 NTUs, the meter generates an accurate turbidity reading in seconds.
  • Infrared LED measurement
  • No maintenance
  • IP 67 (waterproof) rated
  • EPA- approved standards
  • Three years warranty
Generate an accurate turbidity reading quickly with the VWR® Turbidity Meter. Perfect for the plant, field or lab, this simple yet powerful turbidity meter offers an impressive array of features. Use the Infrared (IR) LED light source to comply with ISO 7027 requirements and reliably measure turbidity with auto-ranging values from 0 to 1000 NTU. This meter offers a long battery life averaging about 3000 tests.
Certifications: TUV 3-in-1, ISO 7027.
Delivery information: Kit includes batteries, vials, turbidity standards, silicone oil and cloth, and hard sided carrying case.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty