Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Flex Comfort Waist EMT Pants, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

LP2377BK4030 LP2377BK48UL LP2377MD4234 LP2377MD40UL LP2377BK3832 LP2377MD42UL LP2377BK3830 LP2377BK3432 LP2377MD4832 LP2377MD3030 LP2377MD4834 LP2377MD4634 LP2377MD3032 LP2377MD38UL LP2377BK4430 LP2377MD36UL LP2377BK3632 LP2377MD3430 LP2377MD4030 LP2377MD4432 LP2377MD34UL LP2377BK38UL LP2377BK3032 LP2377BK3434 LP2377MD3034 LP2377BK4632 LP2377BK3834 LP2377BK4234 LP2377MD4230 LP2377MD3232 LP2377MD46UL LP2377BK40UL LP2377BK4830 LP2377MD4232 LP2377BK44UL LP2377MD3234 LP2377BK3630 LP2377BK3030 LP2377MD32UL LP2377MD4632 LP2377MD3434 LP2377BK4032 LP2377MD4032 LP2377BK3232 LP2377BK42UL LP2377MD44UL LP2377BK4434 LP2377BK4630 LP2377MD3630 LP2377BK4832 LP2377MD3432 LP2377MD3830 LP2377MD3834 LP2377BK4634 LP2377BK3234 LP2377BK4232 LP2377MD4034 LP2377MD4630 LP2377BK4230 LP2377BK3230 LP2377BK4834 LP2377MD4434 LP2377MD3230 LP2377BK46UL LP2377MD30UL LP2377BK4432 LP2377BK36UL LP2377BK4034 LP2377MD4430 LP2377BK3634 LP2377BK3430 LP2377MD3634 LP2377MD48UL LP2377MD3632 LP2377BK3034 LP2377MD3832 LP2377MD4830 LP2377BK30UL LP2377BK32UL LP2377BK34UL
76152-190EA 129.32 USD InStock
76152-190 76152-194 76152-150 76152-192 76153-158 76152-118 76152-116 76152-198 76152-154 76152-110 76152-152 76152-196 76152-114 76152-158 76152-156 76152-112 76147-698 76152-162 76147-694 76152-160 76147-696 76153-162 76147-692 76153-164 76153-166 76153-168 76152-206 76152-128 76152-204 76153-160 76152-208 76152-122 76152-166 76152-120 76152-164 76152-126 76152-202 76152-168 76152-124 76152-200 76147-700 76152-096 76152-172 76152-094 76152-170 76153-174 76153-176 76152-138 76153-170 76153-172 76152-210 76152-176 76152-132 76152-098 76152-174 76152-130 76152-136 76152-212 76152-134 76152-178 76153-666 76152-180 76153-668 76152-184 76152-140 76152-182 76153-662 76153-664 76152-108 76152-106 76152-100 76152-188 76152-144 76152-186 76152-142 76152-148 76152-104 76147-702 76152-102 76152-146
Flex Comfort Waist EMT Pants, Dickies®
Workwear
Dickies® style LP2377.

  • StayDark® technology, triple grip waistband
  • Double hook and eye closure, reinforced double knee
  • Full size functional cargo pockets
  • Tool pocket with heavy-duty snap loops
  • Heavy-duty brass ratcheting zipper
  • 7.25 oz. stretch twill fabric, 65% polyester/35% cotton

Sizing Chart: Size measurements indicate the waist by inseam, in inches. UL size designations represent a 39" inseam, unhemmed.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty