Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Industrial 100% Cotton Cargo Pants, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

LP337BK3634 LP337CH58UU LP337DS3632 LP337NV36UL LP337CH3034 LP337NV52UU LP337CH46UU LP337BK4034 LP337CH50UU LP337CH3634 LP337CH3432 LP337BK3630 LP337NV48UU LP337BK3034 LP337CH3832 LP337NV3030 LP337BK52UU LP337CH38UL LP337CH3232 LP337DS42UU LP337NV3232 LP337CH3434 LP337DS3634 LP337DS46UU LP337BK30UL LP337BK3234 LP337DS54UU LP337CH30UL LP337NV4232 LP337NV3630 LP337BK34UL LP337CH4032 LP337BK3434 LP337CH48UU LP337CH42UU LP337DS60UU LP337DS4234 LP337BK3632 LP337CH4030 LP337NV46UU LP337BK50UU LP337DS3630 LP337CH29UU LP337DS3830 LP337NV3034 LP337BK29UU LP337DS34UL LP337NV40UU LP337NV34UL LP337CH4232 LP337DS3234 LP337CH3632 LP337DS3832 LP337DS4032 LP337DS3230 LP337NV60UU LP337NV31UL LP337NV58UU LP337BK3232 LP337BK4230 LP337NV4034 LP337DS31UL LP337CH4230 LP337CH3630 LP337NV44UU LP337DS29UU LP337BK3030 LP337BK3830 LP337BK3832 LP337DS44UU LP337DS4030 LP337NV3430 LP337BK56UU LP337DS58UU LP337BK3834 LP337NV30UL LP337CH34UL LP337NV3834 LP337BK33UL LP337CH52UU LP337NV35UL LP337DS4034 LP337CH36UL LP337BK38UL LP337NV3632 LP337NV54UU LP337BK40UU LP337DS3432 LP337DS3030 LP337CH3830 LP337CH56UU LP337NV56UU LP337NV42UU LP337BK28UU LP337BK60UU LP337CH4034 LP337DS3034 LP337DS3430 LP337DS3834 LP337NV4230 LP337CH54UU LP337DS4232 LP337NV4234 LP337NV38UL LP337CH3230 LP337CH3032 LP337DS4230 LP337CH3430 LP337BK35UL LP337CH31UL LP337NV3830 LP337BK4032 LP337NV3032 LP337NV3434 LP337DS52UU LP337DS28UU LP337NV32UL LP337BK4232 LP337DS3434 LP337NV4032 LP337BK3032 LP337CH40UU LP337DS30UL LP337BK44UU LP337CH28UU LP337DS40UU LP337BK46UU LP337DS48UU LP337DS36UL LP337BK3430 LP337NV3432 LP337NV3832 LP337DS35UL LP337CH3234 LP337BK58UU LP337NV3634 LP337BK32UL LP337NV3230 LP337BK31UL LP337CH4234 LP337BK36UL LP337CH3030 LP337BK48UU LP337DS3232 LP337NV3234 LP337BK4234 LP337NV28UU LP337DS33UL LP337BK42UU LP337CH60UU LP337BK3230 LP337NV4030 LP337CH32UL LP337CH33UL LP337NV29UU LP337DS38UL LP337CH35UL LP337BK54UU LP337NV50UU LP337DS3032 LP337DS56UU LP337DS50UU LP337BK3432 LP337BK4030 LP337DS32UL LP337CH44UU LP337CH3834 LP337NV33UL
76153-676EA 88.33 USD InStock
76153-676 76153-678 76152-470 76153-670 76153-672 76153-674 76152-238 76152-358 76152-232 76152-474 76152-352 76152-472 76152-230 76152-350 76152-478 76152-236 76152-356 76152-476 76152-234 76152-354 76153-688 76153-204 76153-206 76152-360 76153-208 76152-480 76153-680 76153-682 76153-200 76153-684 76153-686 76153-202 76152-404 76152-248 76152-402 76152-408 76152-406 76152-364 76152-242 76152-362 76152-240 76152-368 76152-246 76152-400 76152-366 76152-244 76153-698 76152-250 76152-370 76153-690 76153-692 76153-178 76153-694 76153-210 76153-212 76153-696 76152-416 76152-414 76152-418 76152-254 76152-374 76152-252 76152-372 76152-258 76152-412 76152-378 76152-410 76152-256 76152-376 76152-382 76152-260 76152-380 76153-184 76153-186 76153-188 76152-306 76152-426 76152-304 76152-424 76153-180 76152-308 76153-182 76152-428 76152-386 76152-264 76152-384 76152-262 76152-302 76152-422 76152-268 76152-388 76152-300 76152-266 76152-420 76152-272 76152-392 76152-270 76152-390 76153-196 76153-198 76152-438 76152-316 76152-436 76153-190 76152-314 76153-192 76153-194 76152-318 76152-430 76152-276 76152-396 76152-274 76152-394 76152-434 76152-312 76152-278 76152-432 76152-398 76152-310 76152-282 76152-280 76152-328 76152-448 76152-326 76152-446 76152-320 76152-440 76152-286 76152-284 76152-324 76152-444 76152-322 76152-288 76152-442 76152-290 76152-294 76152-292 76152-218 76152-338 76152-458 76152-216 76152-336 76152-298 76152-452 76152-330 76152-296 76152-450 76152-456 76152-214 76152-334 76152-454 76152-332 76153-700 76152-228 76152-348 76152-468 76152-226 76152-342 76152-220 76152-462 76152-340 76152-460 76152-346 76152-224 76152-466 76152-344 76152-222 76152-464
Industrial 100% Cotton Cargo Pants, Dickies®
Workwear
Dickies® style LP337.

  • Fade resistant, wrinkle resistant, enhanced durability
  • Wider, stronger belt loops
  • Stitched down cargo pocket flaps
  • Two roomy cargo pockets with hidden zipper closure
  • Extra pocket on outside of cargo pockets
  • Metal tack button waist closure
  • 8.5 oz. vat dyed twill, 100% cotton

Sizing chart: Size measurements indicate the waist by inseam, in inches. UU size designations represent a 37" inseam, unhemmed. UL size designations represent a 39" inseam, unhemmed.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty