Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tactical Relaxed Fit Ripstop Cargo Pants, Dickies®

Supplier: Dickies

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

LP703DS4034 LP703DS30UL LP703MD40UL LP703BK3632 LP703BK3830 LP703BK44UL LP703BK40UL LP703BK4032 LP703BK4034 LP703MD4232 LP703BK4434 LP703DS4234 LP703BK42UL LP703DS38UL LP703DS3634 LP703MD3434 LP703MD4634 LP703MD48UL LP703BK3232 LP703BK4232 LP703MD3430 LP703MD34UL LP703MD36UL LP703MD4032 LP703DS4830 LP703BK4832 LP703BK3630 LP703DS4432 LP703DS3034 LP703BK4230 LP703DS48UL LP703BK3234 LP703DS4430 LP703DS32UL LP703MD3030 LP703DS3830 LP703BK4632 LP703MD3632 LP703DS3632 LP703MD4434 LP703DS34UL LP703BK36UL LP703MD4834 LP703MD3032 LP703BK4430 LP703BK4830 LP703BK4432 LP703MD4632 LP703DS3834 LP703BK3230 LP703MD4234 LP703DS3030 LP703BK4630 LP703MD38UL LP703BK3834 LP703DS44UL LP703DS4434 LP703MD3232 LP703BK4834 LP703DS3232 LP703MD3630 LP703MD42UL LP703MD3832 LP703BK4030 LP703BK46UL LP703BK34UL LP703MD32UL LP703MD3234 LP703MD44UL LP703DS4634 LP703BK3030 LP703BK3434 LP703MD3034 LP703MD46UL LP703MD3230 LP703BK3634 LP703MD3634 LP703BK4634 LP703DS4030 LP703DS3832 LP703DS3032 LP703DS4630 LP703MD30UL LP703DS4032 LP703MD4230 LP703BK32UL LP703BK4234 LP703MD4430 LP703DS3230 LP703DS4232 LP703MD4630 LP703DS4230 LP703DS36UL LP703MD4034 LP703MD4432 LP703DS4632 LP703MD4832 LP703MD4830 LP703DS40UL LP703BK3832 LP703BK3032 LP703BK3432 LP703BK3034 LP703DS3434 LP703DS4832 LP703MD4030 LP703MD3830 LP703MD3834 LP703DS3630 LP703DS42UL LP703BK3430 LP703MD3432 LP703DS4834 LP703BK38UL LP703DS3432 LP703DS46UL LP703BK48UL LP703BK30UL LP703DS3234 LP703DS3430
76153-798EA 154.57 USD InStock
76153-798 76153-830 76153-832 76153-834 76153-836 76153-790 76153-792 76153-794 76153-796 76153-828 76154-008 76153-842 76153-844 76153-846 76153-328 76153-848 76153-726 76153-840 76153-838 76153-732 76153-852 76153-610 76153-734 76153-854 76153-612 76153-736 76153-856 76153-614 76153-738 76153-858 76153-616 76153-330 76153-332 76153-730 76153-334 76153-850 76154-022 76154-020 76153-608 76153-728 76153-622 76153-864 76153-742 76153-866 76153-624 76153-744 76153-868 76153-626 76153-746 76153-628 76153-748 76153-860 76153-620 76153-862 76153-740 76154-014 76154-012 76154-010 76153-618 76154-018 76154-016 76153-754 76153-874 76153-632 76153-756 76153-634 76153-876 76153-758 76153-636 76153-878 76153-638 76153-750 76153-870 76153-752 76153-872 76153-630 76153-886 76153-644 76153-764 76153-646 76153-800 76153-766 76153-648 76153-802 76153-768 76153-804 76153-880 76153-640 76153-882 76153-760 76153-642 76153-884 76153-762 76153-776 76153-654 76153-778 76153-810 76153-656 76153-658 76153-812 76153-814 76153-770 76153-772 76153-650 76153-774 76153-652 76153-806 76153-808 76153-820 76153-786 76153-822 76153-788 76153-824 76153-826 76153-660 76153-780 76153-782 76153-784 76153-816 76153-818
Tactical Relaxed Fit Ripstop Cargo Pants, Dickies®
Workwear
Dickies® style LP703.

  • Dupont Tephlon® fabric protector repels water and stains
  • Comfort waist, tunnel belt loops
  • Cell phone pocket
  • Roomy bellowed cargo pockets with built-in mag pouch
  • L-shaped front pocket for knife, double knee with vent for knee pad
  • Triple waistband gripper, double seat for added durability
  • Deep back pockets with additional welt pockets

Features double reinforced inside pocket bags and made of 6.5 oz. ripstop, 65% polyester/35% cotton.

Sizing chart: Size measurements indicate the waist by inseam, in inches. UL size designations represent a 39" inseam, unhemmed.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty