Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Basic Industrial Comfort Waist Pants, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

LP817BK60UU LP817NV48UU LP817DS33UL LP817NV29UU LP817CH60UU LP817NV30UL LP817DS30UL LP817DS42UU LP817BK42UU LP817DS40UU LP817NV42UU LP817NV52UU LP817CH28UU LP817BK50UU LP817CH50UU LP817DS38UL LP817DS48UU LP817CH58UU LP817BK29UU LP817NV58UU LP817CH44UU LP817DS52UU LP817DS36UL LP817CH38UL LP817DS34UL LP817BK33UL LP817NV33UL LP817NV46UU LP817CH30UL LP817DS50UU LP817CH54UU LP817NV34UL LP817DS54UU LP817CH56UU LP817BK48UU LP817CH46UU LP817DS44UU LP817DS35UL LP817BK46UU LP817NV44UU LP817CH36UL LP817DS29UU LP817BK32UL LP817CH31UL LP817BK34UL LP817BK30UL LP817DS60UU LP817BK52UU LP817CH33UL LP817NV28UU LP817NV54UU LP817BK44UU LP817CH52UU LP817DS56UU LP817BK56UU LP817BK40UU LP817NV50UU LP817NV40UU LP817NV56UU LP817CH32UL LP817BK35UL LP817CH48UU LP817BK31UL LP817CH40UU LP817CH42UU LP817CH35UL LP817BK36UL LP817DS32UL LP817BK58UU LP817BK38UL LP817BK54UU LP817CH29UU LP817BK28UU LP817NV35UL LP817DS31UL LP817NV60UU LP817NV32UL LP817NV31UL LP817NV36UL LP817CH34UL LP817DS46UU LP817NV38UL LP817DS58UU LP817DS28UU
76154-918EA 72.57 USD InStock
76154-918 76154-916 76154-838 76154-914 76154-640 76154-882 76154-880 76154-924 76154-648 76154-802 76154-846 76154-922 76154-646 76154-888 76154-844 76154-800 76154-920 76154-886 76154-842 76154-644 76154-840 76154-642 76154-884 76154-908 76158-268 76154-906 76154-828 76154-904 76154-826 76154-872 76154-794 76154-870 76154-792 76154-790 76154-638 76154-836 76154-912 76154-878 76154-834 76154-910 76154-876 76154-832 76154-798 76154-874 76154-830 76154-796 76154-818 76154-816 76158-274 76158-272 76158-270 76154-860 76154-902 76154-868 76154-824 76154-900 76154-822 76154-866 76154-788 76154-820 76154-864 76154-862 76154-808 76154-806 76154-804 76154-848 76154-850 76154-894 76154-652 76154-892 76154-650 76154-890 76154-814 76154-858 76154-856 76154-658 76154-812 76154-854 76154-810 76154-898 76154-656 76154-852 76154-654 76154-896
Basic Industrial Comfort Waist Pants, Dickies®
Workwear
Dickies® style LP817.

  • Stay Dark® technology, wrinkle resistant, stain release
  • Enhanced durability, wider and stronger belt loops
  • Hidden expandable comfort waistband, button closure
  • Reinforced, deep front pockets
  • Left back pocket has button closure
  • 7.75 oz. vat dyed twill, 65% polyester/35% cotton

Sizing Chart: UU size designations represent a 37" inseam, unhemmed. UL size designations represent a 39" inseam, unhemmed.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty