Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Industrial 11" Flat Front Shorts, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

LR303NV32 LR303DS32 LR303NV34 LR303DS38 LR303BK35 LR303NV31 LR303BK36 LR303DS35 LR303NV50 LR303BK30 LR303BK48 LR303NV28 LR303NV40 LR303BK33 LR303BK40 LR303NV35 LR303NV38 LR303BK34 LR303NV33 LR303BK31 LR303BK42 LR303DS34 LR303NV36 LR303BK38 LR303NV30 LR303BK28 LR303NV48 LR303DS48 LR303BK29 LR303DS31 LR303DS46 LR303NV29 LR303BK46 LR303BK50 LR303NV44 LR303DS36 LR303DS40 LR303NV46 LR303NV42 LR303BK44 LR303DS44 LR303DS30 LR303DS42 LR303BK32 LR303DS29 LR303DS50 LR303DS28 LR303DS33
76155-286EA 56.78 USD InStock
76155-286 76155-242 76155-264 76155-288 76155-244 76155-266 76155-222 76155-282 76155-260 76155-262 76158-298 76155-240 76155-284 76155-228 76158-296 76155-304 76158-294 76155-306 76155-268 76158-292 76155-224 76155-246 76155-300 76155-226 76155-248 76155-302 76155-280 76155-296 76155-252 76155-274 76155-230 76155-298 76155-232 76155-276 76155-254 76155-292 76155-270 76155-272 76155-250 76155-294 76155-238 76155-234 76155-278 76155-256 76155-236 76155-258 76155-308 76155-290
Industrial 11" Flat Front Shorts, Dickies®
Workwear
Dickies® style LR303.

  • StayDark® technology, resists wrinkles, easy care stain release
  • Enhanced durability, wider and stronger belt loops
  • Reinforced, deep front pockets
  • Left back pocket has button closure
  • 7.75 oz. vat dyed twill, 65% polyester/35% cotton

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty