Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Industrial 11" 100% Cotton Cargo Shorts, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

LR337BK35 LR337DS31 LR337BK34 LR337DS34 LR337NV36 LR337NV42 LR337BK42 LR337NV40 LR337DS44 LR337NV38 LR337DS48 LR337BK33 LR337DS28 LR337BK32 LR337DS35 LR337DS36 LR337NV33 LR337BK38 LR337NV30 LR337DS33 LR337BK46 LR337BK30 LR337DS30 LR337NV28 LR337NV35 LR337BK31 LR337NV46 LR337BK48 LR337BK50 LR337DS50 LR337NV29 LR337DS42 LR337NV34 LR337NV32 LR337NV50 LR337NV44 LR337NV31 LR337DS46 LR337NV48 LR337BK40 LR337BK44 LR337BK29 LR337DS32 LR337BK28 LR337DS29 LR337DS40 LR337BK36 LR337DS38
76155-440EA 78.84 USD InStock
76155-440 76155-384 76155-340 76155-362 76155-442 76155-386 76155-320 76155-342 76155-364 76155-380 76155-382 76155-360 76155-348 76155-326 76155-328 76155-444 76155-322 76155-366 76155-344 76155-324 76155-346 76155-368 76158-306 76158-304 76155-352 76155-374 76155-330 76155-354 76155-332 76155-376 76158-302 76155-310 76155-370 76158-300 76155-372 76155-350 76155-316 76155-338 76155-436 76155-318 76155-438 76155-356 76155-378 76155-334 76155-312 76155-358 76155-336 76155-314
Industrial 11" 100% Cotton Cargo Shorts, Dickies®
Workwear
Dickies® style LR337.

  • Fade resistant, resists wrinkles
  • Wider & stronger belt loops
  • Two roomy cargo pockets with hidden zipper closure
  • Extra pocket on outside cargo pockets
  • Enhanced durability
  • Metal tack button waist closure
  • Heavy-duty brass ratcheting zipper
  • 8.5 oz. vat dyed twill, 100% cotton

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty