Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Industrial 11" Relaxed Fit Multi-Use Pocket Shorts, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

LR642DC30 LR642NV28 LR642DC33 LR642BK46 LR642BK38 LR642NV48 LR642BK31 LR642BK32 LR642BK40 LR642DC42 LR642DC50 LR642DC28 LR642BK33 LR642DS35 LR642BK35 LR642NV31 LR642NV30 LR642NV44 LR642NV36 LR642BK29 LR642NV46 LR642DS50 LR642DS31 LR642DC35 LR642DS40 LR642NV33 LR642BK48 LR642DC38 LR642BK36 LR642DC34 LR642DS44 LR642DS38 LR642DC36 LR642DC46 LR642DS30 LR642BK30 LR642DC44 LR642NV50 LR642BK28 LR642NV34 LR642DS32 LR642BK44 LR642DC32 LR642BK34 LR642NV35 LR642BK50 LR642DS29 LR642DC40 LR642DS48 LR642BK42 LR642NV40 LR642DC29 LR642NV42 LR642NV38 LR642NV32 LR642DC48 LR642DS46 LR642DS33 LR642DS28 LR642NV29 LR642DC31 LR642DS36 LR642DS34 LR642DS42
76155-682EA 63.08 USD InStock
76155-682 76155-640 76155-684 76155-680 76155-602 76155-646 76155-604 76155-648 76155-400 76155-642 76155-600 76155-402 76155-644 76155-638 76155-594 76155-396 76155-670 76155-398 76155-596 76155-672 76155-392 76155-590 76155-394 76155-592 76155-678 76155-634 76155-636 76155-598 76155-630 76155-674 76155-676 76155-632 76155-628 76155-390 76155-660 76155-582 76155-662 76155-584 76155-580 76155-668 76155-624 76155-626 76155-620 76155-664 76155-388 76155-586 76155-666 76155-622 76155-588 76155-616 76155-618 76155-574 76155-650 76155-612 76155-656 76155-658 76155-614 76155-576 76155-652 76155-578 76155-654 76155-610 76155-606 76155-608
Industrial 11" Relaxed Fit Multi-Use Pocket Shorts, Dickies®
Workwear
Dickies® style LR642.

  • StayDark® technology, enhanced durability
  • Permanent Press post-cured finish, metal tack button waist closure
  • Crotch gusset, wider and stronger belt loops
  • Reinforced front pockets
  • Multi-use side pocket with hidden snap closure

Made of 7.75 oz. vat dyed twill, 65% polyester and 35% cotton.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty