Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Performance Polo without Pocket, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

LS405WH3X LS405WH4X LS405WH2X LS405BK5X LS405BKL LS405WHM LS405DCM LS405ER2X LS405ERL LS405DN4X LS405ER4X LS405DCXL LS405WHL LS405DC2X LS405BKS LS405ER5X LS405DNL LS405DC3X LS405BK3X LS405ER3X LS405DN3X LS405WH5X LS405WHXL LS405DN2X LS405BKM LS405WHS LS405BK2X LS405DN5X LS405DNXL LS405ERXL LS405ERM LS405DC5X LS405BKXL LS405BK4X LS405DNS LS405DCS LS405DCL LS405DNM LS405ERS LS405DC4X
76155-880EA 63.08 USD InStock
76155-880 76158-358 76155-860 76155-882 76158-356 76158-354 76158-352 76155-866 76155-888 76158-350 76155-868 76155-900 76155-862 76155-884 76155-886 76155-864 76155-858 76158-348 76155-890 76158-346 76155-870 76155-892 76158-344 76158-342 76158-340 76158-362 76155-898 76155-854 76155-876 76158-360 76155-878 76155-856 76155-894 76155-872 76155-896 76155-874 76155-906 76155-908 76155-902 76155-904
Performance Polo without Pocket, Dickies®
Workwear
Dickies® style LS405.

  • Resists wrinkles, easy care stain release
  • Color retention wash after wash
  • Snag resistant and requires less dry time
  • Dyed to match buttons
  • Self-collar, side vents, drop tail

Made of 6 oz. micro pique, 100% polyester.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty