Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Industrial Carpenter Jeans, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

LU200RNB4034 LU200RNB4232 LU200RNB44UL LU200RNB3830 LU200RNB3832 LU200RNB3232 LU200RNB50UL LU200RNB4430 LU200RNB3834 LU200RNB4036 LU200RNB36UL LU200RNB3836 LU200RNB3032 LU200RNB4230 LU200RNB58UL LU200RNB3236 LU200RNB3034 LU200RNB32UL LU200RNB3436 LU200RNB3630 LU200RNB4032 LU200RNB46UL LU200RNB4434 LU200RNB5030 LU200RNB4632 LU200RNB3432 LU200RNB42UL LU200RNB30UL LU200RNB3332 LU200RNB5032 LU200RNB52UL LU200RNB3030 LU200RNB4030 LU200RNB28UL LU200RNB3334 LU200RNB40UL LU200RNB3632 LU200RNB3634 LU200RNB3636 LU200RNB3434 LU200RNB54UL LU200RNB38UL LU200RNB4830 LU200RNB3330 LU200RNB33UL LU200RNB4432 LU200RNB34UL LU200RNB56UL LU200RNB31UL LU200RNB4630 LU200RNB4234 LU200RNB4832 LU200RNB3234 LU200RNB48UL LU200RNB3430 LU200RNB60UL LU200RNB3230
76156-728EA 72.57 USD InStock
76156-728 76156-726 76159-614 76159-612 76156-690 76156-694 76159-384 76156-692 76156-698 76156-730 76159-380 76156-696 76159-382 76156-734 76156-732 76156-738 76156-736 76159-608 76159-606 76159-604 76159-602 76159-610 76159-378 76159-374 76159-376 76156-742 76159-370 76159-372 76156-740 76156-746 76156-702 76156-744 76156-700 76156-706 76156-704 76156-708 76160-050 76159-366 76159-600 76159-368 76159-364 76156-712 76156-710 76156-716 76156-714 76156-718 76160-044 76160-048 76160-046 76156-682 76156-680 76156-720 76156-686 76156-684 76156-724 76156-722 76156-688
Industrial Carpenter Jeans, Dickies®
Workwear
Dickies® style LU200.

  • Designed for industrial laundries
  • Triple needle stitched side seams yoke and seat seams
  • Heavy-duty ratcheting brass zipper
  • Hidden loop, dual tool pockets
  • 13.75 oz. premium denim, 100% cotton

Sizing chart: Size measurements indicate the waist by inseam, in inches. UL size designations represent a 39" inseam, unhemmed.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty