Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tactical 10" Relaxed Fit Stretch Ripstop Cargo Shorts

Supplier: Dickies

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

LR704DS36 LR704BK30 LR704BK34 LR704BK42 LR704DS34 LR704GC48 LR704DS30 LR704BK40 LR704BK36 LR704GC46 LR704GC38 LR704MD36 LR704BK46 LR704MD30 LR704MD44 LR704GC36 LR704GC40 LR704DS38 LR704DS40 LR704BK32 LR704MD38 LR704GC30 LR704GC34 LR704MD32 LR704GC42 LR704MD42 LR704MD50 LR704DS46 LR704GC32 LR704MD48 LR704DS44 LR704GC50 LR704MD40 LR704DS48 LR704GC44 LR704DS42 LR704DS32 LR704BK38 LR704MD46 LR704DS50 LR704BK48 LR704MD34 LR704BK44 LR704BK50
76158-314EA 123.02 USD InStock
76158-314 76158-312 76155-760 76158-310 76155-744 76155-722 76155-700 76155-702 76155-746 76155-724 76155-686 76155-740 76155-762 76155-688 76155-764 76155-742 76155-720 76155-738 76155-716 76155-718 76158-308 76155-692 76155-750 76155-694 76155-690 76155-734 76155-756 76155-712 76155-758 76155-714 76155-736 76155-730 76155-752 76155-696 76155-732 76155-710 76155-754 76155-698 76155-708 76155-726 76155-704 76155-748 76155-728 76155-706
Tactical 10" Relaxed Fit Stretch Ripstop Cargo Shorts
Workwear
Dickies® style LR704.

  • L-shaped front pocket to clip a knife
  • Roomy cargo pockets with built in mag pouch
  • Cell phone pocket and inset back pockets
  • DuPont Teflon® finish repels water and stains
  • Sorona fabric provides superior comfort

Dickies® has delivered exceptional performance at unrivaled value, and the legacy continues with field-tested, purpose-built tactical wear. 6.5 oz. ripstop fabric, 43% elasterell-P with sorona/34% cotton/23% polyester. Expandable comfort waist with triple gripper waistband with covert handcuff pocket at center waist.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty