Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Premium 100% Cotton Flat Front Pants, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

WP314KH34UU WP314KH3230 WP314KH3630 WP314KH3830 WP314BK4032 WP314KH3030 WP314KH3634 WP314BK3032 WP314DN3030 WP314DN38UU WP314KH36UU WP314BK3330 WP314DN2832 WP314BK3834 WP314DN3834 WP314DN34UU WP314KH3232 WP314KH3430 WP314BK4430 WP314BK3432 WP314DN4232 WP314BK3232 WP314DN3632 WP314KH2832 WP314DN4432 WP314DN32UU WP314DN3332 WP314DN3430 WP314BK38UU WP314BK4232 WP314BK32UU WP314KH38UU WP314KH4432 WP314KH4230 WP314DN40UU WP314BK4230 WP314BK3630 WP314KH4032 WP314KH3432 WP314DN4430 WP314DN3434 WP314KH3234 WP314DN4832 WP314BK3434 WP314DN3330 WP314BK3634 WP314DN4030 WP314DN3032 WP314KH3834 WP314BK4832 WP314BK34UU WP314BK40UU WP314DN4032 WP314KH3434 WP314KH4430 WP314DN3232 WP314BK3030 WP314DN4632 WP314KH3330 WP314DN3832 WP314DN3230 WP314BK4632 WP314KH3032 WP314BK3632 WP314BK2832 WP314KH3832 WP314BK3230 WP314BK4030 WP314BK3832 WP314KH4232 WP314BK3430 WP314DN36UU WP314DN3630 WP314KH40UU WP314KH4832 WP314KH3632 WP314KH4632 WP314DN3432 WP314DN3634 WP314BK36UU WP314BK3830 WP314DN3234 WP314DN3830 WP314DN4230 WP314BK4432 WP314KH3332 WP314KH32UU WP314KH4030 WP314BK3234 WP314BK3332
76161-278EA 69.41 USD InStock
76161-278 76161-198 76161-276 76160-190 76160-194 76160-270 76160-192 76160-198 76160-230 76160-274 76160-196 76160-272 76160-312 76160-278 76160-234 76160-310 76160-276 76160-232 76160-238 76160-236 76161-192 76161-190 76161-196 76161-194 76161-188 76160-260 76160-182 76160-264 76160-220 76160-186 76160-262 76160-184 76160-268 76160-224 76160-300 76160-266 76160-222 76160-188 76160-228 76160-304 76160-226 76160-302 76160-308 76160-306 76161-212 76161-210 76161-216 76161-214 76160-172 76160-292 76160-290 76161-218 76160-176 76160-296 76160-252 76160-174 76160-294 76160-250 76160-212 76160-256 76160-178 76160-210 76160-254 76160-298 76160-216 76160-214 76160-258 76160-218 76161-200 76161-204 76161-202 76161-208 76160-282 76160-280 76161-206 76160-242 76160-286 76160-240 76160-284 76160-202 76160-246 76160-288 76160-244 76160-200 76160-206 76160-204 76160-248 76161-280 76160-208 76161-282
Premium 100% Cotton Flat Front Pants, Dickies®
Workwear
Dickies® style WP314.

  • Classic cotton work pants
  • Durable 8 oz. twill fabric, 100% cotton
  • Casual waistband
  • Wrinkle resistant
  • Stain release finish means they look great wash after wash
  • Relaxed fit, sits slightly below waist, flat front

Sizing chart: Size measurements indicate the waist by inseam, in inches. UU size designations represent a 37" inseam, unhemmed.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty