Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Relaxed Straight Fit 5-Pocket Denim Jeans, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

13292RBB4232 13293SNB3832 13293THK3634 13292RBB3232 13293SNB3430 13293THK3130 13292RBB4036 13293SNB4632 13292RBB3632 13293RNB3334 13293SNB3636 13293SNB4032 13293THK3030 13292RBB3030 13293RNB3630 13293SNB3434 13293RNB4230 13293RNB4034 13293SNB5030 13293THK3436 13293THK4036 13292RBB3634 13292RBB3432 13292RBB3032 13293RNB3130 13292RBB3834 13292RBB3836 13293THK3832 13293RNB3634 13293THK3636 13293RNB4032 13293RNB5030 13293RNB3032 13293SNB4232 13293SNB4030 13293THK3632 13293THK4030 13292RBB3436 13293THK3334 13293THK4032 13293SNB4036 13293SNB3834 13293SNB3032 13293SNB5032 13293RNB3830 13292RBB4830 13293THK3236 13293RNB3230 13293RNB3832 13293SNB4832 13292RBB4030 13293SNB4034 13293THK5030 13292RBB36UL 13293SNB3230 13293SNB3436 13293SNB3432 13292RBB3332 13293THK4034 13293SNB3836 13293THK4630 13293THK3032 13293RNB4036 13292RBB3334 13292RBB3830 13293SNB3632 13293SNB3034 13293RNB3432 13293SNB4830 13293RNB3834 13292RBB4032 13293RNB4830 13293RNB3232 13293THK3830 13293RNB3332 13293RNB3034 13293RNB3836 13293RNB3430 13293THK3332 13293RNB4432 13293THK5032 13293THK4230 13292RBB4632 13292RBB4432 13292RBB44UL 13293RNB4030 13293THK3132 13293THK3330 13293SNB4630 13292RBB30UL 13293THK4232 13293RNB4630 13293THK4832 13292RBB3430 13293THK3630 13293RNB3636 13293SNB3330 13293THK3434 13293RNB5032 13292RBB3434 13293RNB3330 13293THK3432 13292RBB4832 13293RNB3436 13293SNB4230 13292RBB4630 13293SNB3232 13293SNB3236 13293RNB3234 13292RBB3330 13293THK4830 13292RBB4230 13293SNB3132 13293THK3834 13292RBB3230 13292RBB4034 13293THK4430 13293THK3232 13293THK4632 13293THK3836 13293RNB4430 13293SNB3030 13292RBB40UL 13293RNB3030 13293RNB4832 13293SNB3130 13292RBB3132 13292RBB32UL 13293RNB4232 13292RBB3630 13292RBB42UL 13293THK3430 13292RBB3636 13293SNB3830 13293THK3230 13293RNB3632 13292RBB3234 13293SNB3234 13293THK3234 13293SNB3334 13293SNB3634 13293SNB3630 13292RBB34UL 13292RBB4430 13293RNB3132 13293THK4432 13292RBB38UL 13293RNB3434 13292RBB5030 13292RBB5032 13293SNB4430 13292RBB3832 13293SNB4432 13293RNB3236 13293SNB3332 13293RNB4632 13293THK3034
76161-310EA 75.73 USD InStock
76161-310 76161-314 76161-312 76160-470 76160-592 76160-472 76160-594 76160-590 76160-632 76160-478 76160-512 76160-634 76160-514 76160-474 76160-596 76160-630 76160-476 76160-598 76160-510 76160-636 76160-516 76160-638 76160-518 76161-300 76161-304 76161-302 76160-580 76161-308 76160-460 76160-582 76161-306 76160-462 76160-500 76160-588 76160-622 76160-468 76160-502 76160-624 76160-584 76160-464 76160-586 76160-620 76160-466 76160-508 76160-504 76160-626 76160-506 76160-628 76161-298 76161-732 76161-730 76160-492 76160-494 76161-734 76160-374 76160-490 76160-372 76160-378 76160-534 76160-410 76160-376 76160-536 76160-496 76160-414 76160-530 76160-498 76160-412 76160-532 76161-292 76160-418 76161-290 76160-416 76161-296 76160-538 76161-294 76161-288 76161-286 76160-482 76160-484 76161-728 76160-480 76160-400 76160-522 76160-644 76160-524 76160-646 76160-404 76160-486 76160-640 76160-402 76160-520 76160-488 76160-642 76160-408 76160-406 76160-526 76160-528 76160-392 76160-390 76160-550 76160-396 76160-394 76160-432 76160-556 76160-398 76160-430 76160-558 76160-436 76160-552 76160-554 76160-434 76160-438 76160-380 76160-384 76160-382 76160-544 76160-422 76160-388 76160-546 76160-420 76160-386 76160-426 76160-540 76160-424 76160-542 76160-428 76160-548 76160-570 76160-572 76160-450 76160-610 76160-578 76160-454 76160-612 76160-452 76160-458 76160-574 76160-456 76160-576 76160-618 76160-614 76160-616 76160-560 76160-440 76160-444 76160-566 76160-600 76160-568 76160-442 76160-602 76160-562 76160-448 76160-564 76160-446 76160-608 76160-604 76160-606
Relaxed Straight Fit 5-Pocket Denim Jeans, Dickies®
Workwear
Dickies® styles 13292 and 13293.

  • Brass rivets prevent rips at stress points
  • 7 keystone belt loops set into waistband add strength and prevent bunching
  • Coin pocket fits small pocket knife
  • Signature stitching on back pockets
  • Deep front pockets
  • Heavy-duty brass zipper
  • 100% cotton

The relaxed fit of this traditional jean is roomier through the seat and thigh for comfortable stooping, bending, and kneeling. Slightly tapered, straight leg fits over boots. Constructed of heavyweight denim for years of durable wash-and-wear use.

Sizing chart: Size measurements indicate the waist by inseam, in inches. UL size designations represent a 39" inseam, unhemmed.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty