Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Short Sleeve Heavyweight T-Shirts, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

WS450YGXT WS450DNXT WS450DN5X WS450GH3T WS450BY2X WS450RBLT WS450YG3T WS450RB3T WS450RB2T WS450BVM WS450AG3T WS450FHM WS450ER2T WS450GHL WS450DU1L WS450BVLT WS450HG3X WS450LN3X WS450DSL WS450DU12T WS450HGXL WS450BY5X WS450BKL WS450HG2X WS450CH3X WS450BD2X WS450GH2T WS450WH4X WS450CHS WS450ER3X WS450BVXL WS450CB2T WS450OR3X WS450ERXL WS450FHXT WS450GH2X WS450AGXT WS450DS2T WS450CBXL WS450HGL WS450CBXT WS450DN3X WS450BKLT WS450BKXL WS450HGLT WS450AG3X WS450LNXT WS450BDLT WS450BKXT WS450BYLT WS450RBM WS450FHLT WS450CB3T WS450DNS WS450YG2T WS450WH2T WS450YG3X WS450BK2T WS450DSLT WS450BD4X WS450ORS WS450OR2X WS450WHM WS450BY2T WS450LN3T WS450DNM WS450BDM WS450BKS WS450BDL WS450CHLT WS450FHL WS450CH3T WS450DS3T WS450WH3T WS450HG3T WS450WHL WS450YGXL WS450ERXT WS450YGM WS450ORM WS450ORLT WS450CBM WS450DSM WS450DU13X WS450HG5X WS450DN3T WS450ORXT WS450BDXL WS450BDXT WS450WH2X WS450AG2T WS450DS3X WS450AGM WS450WHS WS450ERL WS450DSS WS450BD2T WS450LNM WS450RBL WS450BV3T WS450CH2T WS450BV2X WS450ERLT WS450OR3T WS450DU1XL WS450DU1XT WS450GHXL WS450DNLT WS450BY4X WS450ORXL WS450BD3T WS450LNL WS450RBXL WS450WH3X WS450WH5X WS450FHXL WS450DN2T WS450OR2T WS450BVXT WS450DU13T WS450YG4X WS450BYXL WS450HG4X WS450YG2X WS450CBLT WS450GHXT WS450GH3X WS450RB2X WS450AGXL WS450BVL WS450ER3T WS450LN2X WS450FH3T WS450BKM WS450ERM WS450FH3X WS450YGL WS450CHL WS450CBL WS450BY3T WS450DU12X WS450WHLT WS450BYXT WS450DNL WS450GHM WS450BK5X WS450LNXL WS450BYL WS450AGLT WS450WHXT WS450BD5X WS450BK3X WS450BK4X WS450DSXL WS450DS4X WS450ER2X WS450CH2X WS450BV3X WS450CHM WS450RB3X WS450DSXT WS450HGXT WS450LNS WS450BK2X WS450WHXL WS450DN4X WS450BV2T WS450BK3T WS450BVS WS450CB3X WS450DU1M WS450CHXT WS450CB2X WS450AGL WS450HGS WS450FH2T WS450YG5X WS450DNXL WS450BY3X WS450HGM WS450YGLT WS450BYM WS450DS2X WS450BD3X WS450DU1LT WS450FH2X WS450CHXL WS450ORL WS450HG2T WS450LNLT WS450LN2T WS450DN2X WS450GHS WS450DS5X WS450GHLT WS450AG2X WS450RBXT
76163-040EA 31.54 USD InStock
76163-040 76163-162 76163-160 76163-048 76163-046 76163-168 76163-044 76163-166 76162-926 76163-042 76163-164 76163-172 76162-928 76163-052 76163-170 76163-050 76162-930 76162-932 76163-178 76162-934 76163-058 76163-176 76163-056 76162-936 76163-174 76162-938 76163-054 76163-260 76163-140 76163-028 76163-268 76163-026 76163-148 76163-024 76163-266 76163-146 76163-264 76163-022 76163-144 76163-262 76163-020 76163-142 76163-150 76163-030 76163-272 76163-270 76163-158 76163-038 76163-156 76163-278 76163-036 76163-154 76163-276 76163-034 76163-152 76163-032 76163-274 76163-084 76163-082 76163-080 76162-960 76162-962 76162-964 76162-966 76162-968 76163-088 76163-086 76163-096 76163-094 76163-092 76163-090 76162-970 76162-972 76162-974 76162-976 76162-978 76163-098 76163-062 76163-184 76163-060 76163-182 76163-180 76162-940 76162-942 76162-944 76163-068 76162-946 76163-066 76163-188 76163-064 76162-948 76163-186 76163-194 76163-074 76163-192 76163-072 76163-190 76163-070 76162-950 76162-952 76162-954 76162-956 76163-198 76163-078 76162-958 76163-196 76163-076 76163-208 76163-206 76163-448 76163-446 76163-204 76163-202 76163-444 76163-200 76163-442 76163-440 76163-218 76163-216 76163-458 76163-456 76163-214 76163-454 76163-212 76163-210 76163-452 76163-450 76162-980 76162-982 76162-984 76162-986 76162-988 76162-990 76162-992 76162-994 76163-438 76163-436 76162-996 76163-434 76162-998 76163-008 76163-006 76163-248 76163-128 76163-246 76163-004 76163-126 76163-002 76163-244 76163-124 76163-000 76163-242 76163-122 76163-240 76163-120 76163-250 76163-138 76163-018 76163-136 76163-258 76163-016 76163-134 76163-014 76163-256 76163-132 76163-012 76163-254 76163-130 76163-010 76163-252 76163-228 76163-108 76163-468 76163-226 76163-106 76163-224 76163-466 76163-104 76163-222 76163-464 76163-102 76163-462 76163-220 76163-100 76163-460 76163-470 76163-118 76163-116 76163-238 76163-114 76163-236 76163-112 76163-234 76163-110 76163-232 76163-472 76163-230
Short Sleeve Heavyweight T-Shirts, Dickies®
Workwear
Dickies® style WS450.

  • Heavyweight pocket tee
  • Tagless back neck label for comfort
  • Taped neck and shoulder seams for longer wear
  • Ash gray: 99% cotton/1% polyester
  • Heather gray: 90% cotton/10% polyester
  • All other colors: 6.75 oz./220 g; 100% cotton

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty