Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Motic BA310 Metallurgical Microscope
Supplier: Motic
1100100403591 1100100402601 1100100402045 1100100402524 1100100402045 1100100402144
76163-628EA 5500.46 USD InStock
76163-628 10767-560 76614-920 10767-558 76163-494 CA10769-624
Motic BA310 Metallurgical Microscope
Microscopes
The Motic BA310Met Advanced Metallurgical microscope series is designed to meet the performance demands of engineering professionals who study opaque samples like minerals, metals, and building materials.
The specialized M Plan achromatic metallurgical objectives (5x, 10x, 20x, 50x) are set in a reversed quintuple nosepiece, providing faithful reproduction of samples up to 30mm in thickness
The BA310Met features a Siedentopf style head inclined at 30° with full 360° rotation for pain-free viewing over extended periods of time. The M Plan Achromatic lenses provide improved working distance (8.2mm at 500x), allowing for sample manipulations while viewing. Additional add-ons such as a polarizer or the Moticam series of microscopy cameras further enhance the BA310Met for metallurgical observation and documentation. The BA310Met provides a robust, reliable tool for education, engineering and material science, industrial quality control, and many other material applications.
The standard halogen epi-illuminator includes adjustable field and aperture diaphragms to minimize reflections and optimize performance. Conversion of the standard halogen illumination to a 3W LED is easily achieved with a simple plug-in module.
Ordering information: Microscope is supplied with all accessories listed above and micrometer reticles. Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty