Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nomex® Flame-Resistant Lab Coat, Women's, CritiCore Protective Wear
Supplier: CritiCore Protective Wear
Nomex®
4950CXP63XL 4950CXP6XXS 4950CXP6L 4950CXP6S 4950CXP6XL 4950CXP6M 4950CXP6XS 4950CXP62XL
76164-548CS 5380.83 USD InStock
76164-548 76163-518 76163-516 76163-514 76163-512 76164-552 76163-520 76164-550
Nomex® Flame-Resistant Lab Coat, Women's, CritiCore Protective Wear
Workwear
Designed to provide a superior level of protection against flash fire, electric arc (HRC 1), and other dangerous conditions, this women's Flame-Resistant (FR) lab coat has numerous applications in both laboratory and production settings. Made with Nomex® CXP6 fabric, it combines exceptional flame resistance with excellent durability, even after repeated industrial launderings.
  • Excellent heat-resistance properties
  • Best in class flammability protection
  • 360 degree FR identification and protection
  • Meets NFPA 70E and HRC 1 performance standards
  • Quick-release snap front
  • Safety sleeve cuff with tunneled elastic
  • Designed to conform with the female shape
Coats feature 360 degree FR identification with patches sewn on the back, plus left and right sleeves
Nomex® is a registered trademark owned by E. I. du Pont de Nemours and Company.
Constructed with a quick-release snap front and safety sleeve cuff with tunneled elastic for additional protection. Coats also feature two lower hip pockets and one breast pocket with pencil slot. Designed specifically for women, this coat is tapered and has a princess-belt back.
Certifications: Meets performance standards for NFPA 2112 and NFPA 70E HRC 1.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty