Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
GM-171 Stereo Microscopes, Motic
Supplier: Motic
1100200600852 1100200600842 1100200600852 1100200600842
76163-544EA 2957.45 USD InStock
CA76163-544 CA76163-542 76163-544 76163-542
GM-171 Stereo Microscopes, Motic
Microscopes
The GM-171 is a sophisticated microscope ready for the demands of jewelers and gem dealers on the examination of gems and precious stones. The magnification is handled by the optical body of Motic’s most recently introduced stereo microscope SMZ-171, available in binocular or trinocular version.
  • Ergonomic setup with clever functionality
  • Microscope stand is tiltable from 0° (upright) to 45° to suit individual seating positions and rotatable by 360° to share visual insights
  • Interpupillary distance can be set from 52 - 75 mm
  • Works perfectly fine with the new 10X widefield eyepieces and the zoom body of 0.75X up to 5X magnification
The GM171 is an advanced solution for jewelry examination, delivering reliable results and adaptable to a wide range of individual requirements.
Ordering information: Contact your Avantor representative to learn more about these and other motic solutions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty