Would you like to log in again?
VWR® Classic Syringe Tips, Non sterile
Supplier: VWR
VWR®
ES40010-0004V ES40010-0003V ES40010-0005V ES40010-0002V ES40007-0009V ES40010-0001V ES40010-0000V ES40007-0010V
75838-210PK 105.5 USD InStock
75838-210 76163-558 76163-598 76163-596 76163-594 75838-208 76163-592 76163-560
VWR® Classic Syringe Tips, Non sterile
Pipette Tips
Classic Syringe Tips provide reliable and repeatable results, and they are compatible with a variety of pipettors.
  • Ideal for use with viscous, infectious, and volatile liquids
  • Compatible with a variety of pipettors
Ripette®, Ripette®pro, Ripette®genX, eLineTM-Dispenser, Minilab 100, Minilab 101, Minilab 201, Distriman®, StepMate, EasyStep, EasyStep electronic, RepeatOne®, Handrop.
These syringe tips are not compatible with: HandyStep®, Repetman®.
Ordering information: The 25 and 50 ml tips require an adapter (needs to be ordered separately).
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty