Would you like to log in again?
Motic BA210E LED Compound Microscopes
Supplier: Motic
1100100403783 1100100403018 1100401200771 1100100403887 1100401200771 1100100403018 1100100403887 1100100403783
76635-284EA 1587 USD InStock
76635-284 CA76163-614 76163-614 CA76635-284 76635-348 CA76635-268 76635-268 CA76635-348
Motic BA210E LED Compound Microscopes
Microscopes
The BA210Elite model introduces superior improvements both in optical and mechanical performance. Meant to be used in Educational life sciences, Medical and a variety of Biological applications, the BA210E is designed for ease of use and longevity due to Motic's complete understanding of the educational market.
  • Widefield high eyepoint eyepiece N-WF10×/20 mm with diopter
  • Adjustment on both eyepieces, with rubber eyecup (paired)
  • Reversed quadruple revolving nosepiece
  • Coaxial coarse and fine focusing adjustment
  • Built in low position coaxial mechanical stage (right-hand control)
  • Focusable Abbe condenser N.A. 1.25 with iris diaphragm and slot
The BA210E model introduces student-proof features both in optical and mechanical aspects. The new rackless stage concept without prominent gear rack allows even more convenient use of the x/y movement, while increasing user safety in teaching situations.
For full freedom of illumination options, the new model BA210E has an easy interchangeability of its 6 V/30 W Halogen bulb with LED modules of different colour temperatures (4500K, 6000K). Extended life-time of LEDs here is combined with cost saving aspects. Light consuming contrast methods like Phase contrast, Polarization or Dark field can easily performed.
Ordering information: Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
Delivery information: The BA210E Phase package option (76163-614) includes upright compound microscope, trinocular head, widefield high eyepoint 10× eyepieces, CCIS EC Plan achromat/phase objectives (PL4×, PL100× S-Oil, Ph10X), rackless stage, NA 1.25 abbe condenser, phase slider, phase centering telescope, 3WLED module.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty