Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Painters Utility Pants, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

1953WH3130 1953WH3432 1953NT3830 1953NT3436 1953WH40UL 1953WH2830 1953WH4030 1953WH3832 1953WH3234 1953WH3436 1953WH34UL 1953NT3430 1953NT3234 1953NT4630 1953WH4430 1953WH4830 1953NT4034 1953NT3432 1953NT3032 1953WH4832 1953NT3834 1953NT3636 1953WH4032 1953NT3030 1953WH38UL 1953NT4632 1953NT3632 1953WH4230 1953NT3232 1953WH3434 1953WH44UL 1953WH3230 1953WH3032 1953WH46UL 1953NT4232 1953NT4030 1953NT3334 1953WH4034 1953NT4430 1953NT4032 1953WH3236 1953WH3430 1953WH3232 1953WH3330 1953WH3636 1953WH3334 1953WH3834 1953WH3134 1953WH4432 1953NT4230 1953WH2930 1953NT3832 1953WH3030 1953WH3836 1953NT4432 1953NT3634 1953NT3332 1953WH4632 1953NT4830 1953NT3034 1953NT3630 1953WH36UL 1953WH3830 1953WH2932 1953WH3132 1953NT3434 1953WH4232 1953WH3632 1953NT3230 1953WH2832 1953NT3330 1953WH3630 1953NT4832 1953WH4234 1953WH3034 1953NT3836 1953WH42UL 1953WH3332 1953WH4630 1953WH3634
76164-724EA 63.08 USD InStock
76164-724 76164-768 76164-846 76164-802 76164-726 76164-804 76164-848 76164-764 76164-842 76164-766 76164-722 76164-844 76164-800 76164-760 76164-762 76164-840 76164-838 76164-812 76164-856 76164-736 76164-814 76164-858 76164-738 76164-852 76164-732 76164-776 76164-854 76164-810 76164-734 76164-778 76164-772 76164-850 76164-774 76164-730 76164-770 76164-728 76164-806 76164-808 76164-746 76164-868 76164-824 76164-748 76164-826 76164-742 76164-786 76164-820 76164-864 76164-744 76164-788 76164-822 76164-866 76164-782 76164-860 76164-740 76164-784 76164-862 76164-780 76164-816 76164-818 76164-834 76164-758 76164-836 76164-874 76164-830 76164-754 76164-798 76164-832 76164-756 76164-870 76164-794 76164-750 76164-872 76164-752 76164-796 76164-790 76164-792 76167-694 76167-698 76164-828 76167-696
Painters Utility Pants, Dickies®
Workwear
Dickies® style 1953.

  • 100% cotton drill fabric and triple-stitched for extra strength
  • Deeper and wider rule pocket on right leg
  • Double tool-and-cell pocket on left leg, with extra-large back pockets
  • Relaxed fit in seat and thigh
  • Slightly tapered leg fits over boots
  • Hammer loops on legs and center back
  • 8.75 oz. painters drill, 100% cotton

Sizing chart: Size measurements indicate the waist by inseam, in inches. UL size designations represent a 39" inseam, unhemmed.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty