Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Heavyweight Cotton Long Sleeve Shirts, Dickies®

Supplier: VF Imagewear
549KH3X 549KHLT 549KHM 549KHS 549KH2X 549KHL 549KHXL 549KHXT
76167-198EA 88.33 USD InStock
76167-198 76165-942 76167-196 76165-940 76165-946 76165-944 76165-938 76165-936
Heavyweight Cotton Long Sleeve Shirts, Dickies®
Workwear
Dickies® style 549.

  • Doghouse sleeve plackets
  • Generous, relaxed fit; 2.5" fuller through chest than Regular fit
  • Casual look
  • Wrinkle resistant
  • Two front chest flap pockets

Chest: 49.5"/Waist: 49.5"/Sweep: 49.5".

Made up of 8.5 oz. twill, 100% cotton.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty