Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Loose Fit Cargo Shorts, 13", Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

43214RBK36 43214RDN40 43214RKH32 43214RKH40 43214RKH44 43214RDN34 43214RKH48 43214RBK32 43214RBK42 43214RDN42 43214RDN30 43214RDN48 43214RBK34 43214RBK46 43214RDN36 43214RBK40 43214RBK44 43214RKH30 43214RKH42 43214RBK48 43214RKH36 43214RKH34 43214RKH46 43214RKH38 43214RDN44 43214RDN38 43214RBK38 43214RBK30 43214RDN32 43214RDN46
76168-042EA 78.84 USD InStock
76168-042 76168-020 76168-044 76168-022 76168-060 76168-040 76168-006 76168-028 76168-008 76168-046 76168-002 76168-024 76168-026 76168-004 76168-048 76168-052 76168-030 76168-032 76168-054 76168-010 76168-050 76168-016 76168-038 76168-018 76168-056 76168-034 76168-012 76168-014 76168-058 76168-036
Loose Fit Cargo Shorts, 13", Dickies®
Workwear
Dickies® style 43214.

  • Loose fit
  • Flat front
  • Comfortable garment washed twill
  • Casual waistband
  • Pleated cargo pockets
  • 8.5 oz. twill, 100% cotton

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty