Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

13" Loose Fit Multi-Use Pocket Work Shorts, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

42283SV30 42283BK34 42283DB44 42283DB50 42283WH36 42283CH30 42283WH30 42283BK40 42283BK28 42283SV29 42283BK31 42283ER52 42283DN28 42283KH56 42283OG40 42283CH33 42283DN54 42283NV54 42283DB38 42283BK36 42283NV40 42283NV29 42283ER56 42283SV33 42283DB32 42283DN42 42283OG32 42283ER29 42283DN34 42283CH28 42283KH58 42283ER34 42283DB28 42283DN60 42283SV31 42283WH29 42283WH32 42283BK33 42283WH50 42283DN32 42283NV30 42283KH38 42283ER42 42283KH36 42283CH54 42283KH52 42283ER36 42283DN50 42283SV44 42283KH46 42283BK30 42283NV46 42283ER50 42283NV28 42283ER38 42283CH46 42283CH34 42283DN46 42283CH29 42283OG36 42283ER28 42283WH40 42283CH60 42283ER48 42283RB32 42283NV44 42283KH30 42283KH32 42283CH48 42283SV38 42283DB42 42283KH54 42283BK42 42283BK44 42283ER60 42283ER54 42283KH44 42283NV48 42283RB44 42283DB29 42283DN40 42283WH33 42283CH50 42283DB40 42283NV32 42283WH31 42283NV31 42283BK52 42283KH50 42283CH42 42283DN56 42283SV42 42283SV50 42283KH40 42283CH56 42283DB33 42283SV28 42283BK48 42283NV50 42283ER33 42283KH33 42283BK54 42283RB36 42283ER46 42283CH31 42283WH44 42283BK46 42283DN52 42283CH40 42283ER58 42283RB30 42283NV36 42283BK29 42283CH44 42283DN44 42283DB34 42283ER32 42283DB31 42283RB42 42283WH28 42283NV52 42283DN33 42283OG34 42283DN29 42283RB34 42283WH42 42283OG38 42283BK50 42283SV34 42283CH36 42283DN36 42283CH52 42283BK60 42283NV42 42283KH34 42283SV32 42283WH48 42283KH29 42283NV33 42283WH34 42283CH58 42283CH32 42283RB38 42283NV34 42283DB48 42283DN48 42283RB40 42283WH46 42283DN38 42283WH38 42283DN30 42283BK32 42283CH38 42283DB36 42283KH28 42283ER31 42283BK38 42283KH31 42283ER40 42283SV40 42283KH48 42283SV46 42283OG30 42283DN31 42283DN58 42283BK58 42283SV36 42283DB46 42283DB30 42283KH60 42283ER30 42283ER44 42283KH42 42283SV48 42283BK56 42283NV38
76167-978EA 63.08 USD InStock
76167-978 76165-608 76165-606 76165-728 76168-540 76165-578 76165-732 76165-612 76167-980 76165-730 76165-576 76165-698 76165-610 76165-736 76168-538 76165-616 76165-734 76165-614 76165-570 76167-988 76165-692 76165-690 76167-986 76165-574 76165-696 76167-984 76165-572 76165-694 76167-982 76167-908 76165-718 76165-716 76165-688 76167-990 76165-600 76165-568 76165-722 76165-686 76165-720 76165-604 76165-726 76165-602 76165-724 76167-998 76165-680 76167-910 76167-996 76165-684 76167-994 76165-682 76167-992 76167-918 76167-916 76167-914 76167-912 76165-708 76165-706 76165-710 76165-678 76165-676 76165-714 76165-712 76167-922 76165-670 76167-920 76165-674 76165-672 76167-928 76167-926 76167-924 76165-666 76165-700 76165-664 76165-704 76165-668 76165-702 76167-932 76165-780 76167-930 76165-662 76165-784 76165-660 76165-782 76167-938 76167-936 76167-934 76168-502 76165-776 76165-656 76165-774 76165-654 76165-778 76165-658 76167-944 76167-942 76165-772 76167-940 76165-652 76165-770 76165-650 76167-948 76167-946 76168-512 76168-514 76168-510 76165-644 76168-508 76165-766 76165-642 76165-764 76168-504 76165-648 76165-646 76168-506 76165-768 76167-954 76167-952 76165-640 76167-950 76165-762 76165-760 76168-000 76167-958 76167-956 76168-522 76168-524 76165-628 76168-520 76165-754 76165-634 76165-598 76165-752 76165-632 76165-758 76168-516 76165-638 76165-756 76168-518 76165-636 76165-592 76167-966 76165-590 76167-964 76165-596 76165-750 76165-630 76167-962 76165-594 76167-960 76167-968 76168-534 76165-738 76165-618 76168-536 76168-530 76168-532 76165-622 76165-744 76165-620 76165-742 76165-588 76165-626 76168-526 76165-748 76168-528 76165-624 76165-746 76167-976 76165-582 76167-974 76165-580 76167-972 76165-586 76165-740 76167-970 76165-584
13" Loose Fit Multi-Use Pocket Work Shorts, Dickies®
Workwear
Dickies® style 42283.

  • Loose fit, flat front
  • Permanent crease
  • Scotchgard™ stain release finish
  • Logo label on left front leg and multi-use side pocket
  • With 13" inseam
  • Made of 8.5 oz. twill, 65% polyester/35% cotton

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty