Would you like to log in again?
Personna Plus® Microtome Blades
Supplier: AccuTec Blades
Personna Plus®
AVBL-2007-0000 AVBL-2005-0000
76169-160CS 916.27 USD InStock
76169-160 76169-162
Personna Plus® Microtome Blades
Microtomes and Microtome Assemblies Blades and Handles
Engineered to perform.
  • Available in High Profile or Low Profile formats
  • Round corners
  • Safety dispenser, used blade vault and round corner blades
  • 3 facet edge
  • Made in U.S.A
There is no room for error when sampling and sectioning. These stainless steel microtome blades are precision ground with evenly matched and aligned blade edges and optimal cutting angles. The propietary MicroCoat® surface produces smoother cuts with less compression and greater section clarity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty