Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cool Offs, Allegro

Supplier: Allegro

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

8405-53 8405-55 8405-51 8405-50 8405-54 8405-59
76169-218EA 3.92 USD InStock
76169-218 76169-222 76169-212 76169-220 76169-216 76169-214
Cool Offs, Allegro
Workwear
Cooling products are designed to protect and provide comfort to workers who must function in environments with excessive heat.

  • Lightweight and comfortable
  • 38" length (longest on the market)
  • 100% cotton

Keep cool all day long with these dual-use Cool Offs

To use, simply soak the garment in cold water for 10 to 15 minutes. The cooling crystals go to work to keep wearers cool for days.

Quilted into four sections for an even distribution of hydration material, Cool Offs can be worn around the neck or tied as a headband. A unique button hole closure ensures a secure fit. The outer shell is 100% cotton and comes in multiple colours and patterns.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty