Would you like to log in again?
VWR® Advanced Sleeves
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
414004-420 414004-421 76169-444 76169-442
414004-421CS 245.9 USD InStock
414004-421 414004-420 76169-442 76169-444
VWR® Advanced Sleeves
Disposable Garments
VWR® Advanced protection sleeves are manufactured from a specially formulated breathable microporous fabric that is exceptionally clean and durable.
  • Significant fluid and particulate barrier
  • Durable, lightweight fabric
  • Excellent breathability and water vapor transmission rate
  • Soft tactile feel
  • Thumb loop versions also available
These protective garments provide a fluid barrier and dry-particulate protection, as well as resistance to common laboratory chemical.
Garments feature sonic-welded seams and serged elastic at both ends. Designed without an attached thumb loop. Sleeves are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty