Would you like to log in again?
ThermoFlex™ Recirculating Chillers, Thermo Scientific
Supplier: Thermo Scientific
1011110000200100 1123020501200200 1011010000200100 1111C10100200100 1011010520200100 1313C10160200200
76169-482EA 8349.58 USD InStock
76169-482 76169-494 76169-480 76169-492 76169-476 76169-478
ThermoFlex™ Recirculating Chillers, Thermo Scientific
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Optimized for the most demanding applications, Thermo Scientific™ ThermoFlex Recirculating Chillers are reliable, easy-to-use and easy-to-maintain.
  • An intuitive user interface for ease of operation
  • Air and water filters that can be changed while unit is in operation
  • Innovative packaging for rapid installation
  • Quick start guide enables trouble-free system start-up
  • Superior performance
  • Improved cooling capacity
  • Configurable design
  • Wide range of available cooling capacities
  • Variety of available options
  • Installation flexibility
The ThermoFlex platform was developed with customer input from concept to design. This high-performance chiller platform, with cooling capacity up to 24,000 watts (81,948 btu/hr), is configurable to meet application demands.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty