Would you like to log in again?
PAD PREMIUM UNIV SORBENT 16 INX18 IN/FT
Supplier: FYTERTECH NONWOVENS LLC
Premium Universal Sorbent Pad Standard Medium-Weight^ Premium Universal Sorbent Pad Standard, Medium-Weight, Dimensions: 16inx18in/ft, Color: Gray, Bonding / Embossing: Dimple, Premium universal sorbents are designed for extended-use applications by utilizing low-lint and high strength meltblown
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PAD PREMIUM UNIV SORBENT 16 INX18 IN/FT 76169-558 GFF-72 Case of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty