Would you like to log in again?
SORBENT ROLL UNIV LOW CAPACITY 32X300IN
Supplier: FYTERTECH NONWOVENS LLC
Universal HiVis Sorbent Roll light Capacity^ Universal HiVis Sorbent Roll light Capacity, Light-Weight, Dimensions: 32inx3000in/ft, Color: Hi-Vis Yellow, Bonding / Embossing: Dimple, multi-use product offering that combines an absorbent with a caution/warning sign, High-Visibility print layer draws
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SORBENT ROLL UNIV LOW CAPACITY 32X300IN 76169-574 HVU-95 Bag of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty