Would you like to log in again?
Pitched Turbine Blade, Caframo
Supplier: Caframo
A561 A561
76169-582EA 130.57 USD InStock
76169-582 CA76147-822
Pitched Turbine Blade, Caframo
Stirrers Stirring Tools
Pitched blade turbine composed of electropolished 316L stainless steel. The 0.8 cm bore is compatible with all Caframo shafts. Axial flow with moderate shear for moderate viscosity mixing.
  • Four-blade pitched turbine
  • Moderate shear axial flow
  • Secured to shaft with set screws
  • Scale up advantage if turbine used in production
  • Serves a wide range of applications
It is chemically inert and corrosion resistant and multiple blades can be added along shaft and easy to clean.
Blade rotation: left hand.
Delivery information: Set screws included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty