Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunc™ Cell Culture EasYDishes, Thermo Scientific
Supplier: Thermo Scientific
Nunc™
150460 150466 150462 150464 150468
76169-590CS 282.38 USD InStock
76169-590 76169-586 76169-584 76169-634 76169-588
Nunc™ Cell Culture EasYDishes, Thermo Scientific
Culture Dishes Cell Culture Dishes
Nunc™ EasYDishes are the next generation of cell culture dishes that improve handling, stacking, and transporting of cell cultures in the lab.
  • Beveled grip ring makes it easier to handle and manage with a gloved hand
  • Proprietary notch design maximizes ventilation while maintaining sterility, and restricts lid movement to ensure secure lid-to-dish fitting
  • Raised outer edge on dish lid allows stable stacking during transportation of cell cultures
  • Orientation marks on dish bottom simplify cell location under the microscope
  • Writing area improves traceability of cell cultures
  • Re-sealable packaging promotes cleanliness of opened packs
Dishes receive the standard Nunclon™ Delta surface treatment to maximize adhesion for the majority of cell types
Nunc™ EasYDishes are available in four sizes; 35 mm, 60 mm, 100 mm, and 150 mm. Cell culture treated and lot-to-lot certified for adhesion cultures.
Reproducibility and reliability are key to good scientific results, which is why every lot of Nunc™ cultureware products is tested with four different cell lines. This ensures unfailing monolayer formation and consistent cloning efficiency.
Certifications: All dishes are gamma irradiated to a Sterility Assurance Level (SAL) of 10-6 and certified RNase/Dnase-free, Human DNA-free, non-cytotoxic, and non-pyrogenic. Dishes come with Certificate of Quality in every case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty