Would you like to log in again?
VWR MINICHILLER 900W OLE 208V IP20 1.25L
Supplier: VWR International
VWR* OLE Chiller 900w 208 VAC 60Hz^ VWR* OLE Chiller 900w 208 VAC 60Hz, Dimensions:23x54x28 cm, Reservoir Capacity: 1.25 L, Max Temperature: 40 deg C, Cooling Capacity At 0 deg C: 700 watts, Max Flow Rate: 25 L/min, Max Pump Pressure: 13.05 PSI, Interface: USB, RS232, Power: 208 VAC 60Hz
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR MINICHILLER 900W OLE 208V IP20 1.25L 76169-616 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty