Would you like to log in again?
Blue Guard Industrial Wipe, High-Tech Conversions
Supplier: High-Tech Conversions
BG-1215
76169-636CS 242.8 USD InStock
76169-636
Blue Guard Industrial Wipe, High-Tech Conversions
Wipes Disposable Wipes and Towels Wipes for General Applications
Heavy-duty, super-tough blue industrial wipe made of poly-cellulose blend.
  • Blue industrial wipe
  • Super-tough and absorbent
Heavy-duty, super-tough blue wipes are absorbent, low-linting, and low in chemical extractables. They are comprised of a blend of absorbant cellulosic fibers and strong polyester fibers. Blue Guard is bulk packed and economically priced.
Packaging: Bulk packed 1000 pieces/case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty