Would you like to log in again?
Accessory Parts for TRIO.BAS™ Air Samplers, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BAS301 BAS522 BAS420 BAS371 BAS524 BAS522 BAS421 BAS524 BAS521 BAS265 BAS523 BAS523 BAS395 BAS520 BAS396 BAS397 BAS521 BAS530 BAS371 BAS197 BAS340 BAS530 BAS331 BAS330 BAS336 BAS380 BAS520 BAS341 BAS465 BAS465 BAS334
75804-700EA 377.67 USD InStock
75804-700 75804-702 75804-746 75804-704 75838-740 75804-748 CA75994-446 CA75994-448 CA75994-452 75994-464 CA75994-454 CA75994-470 75994-468 75994-446 75994-466 CA75994-450 75994-444 76169-672 76509-286 75804-756 75804-758 75994-448 75804-750 75804-696 75804-754 CA75994-464 75994-452 75994-450 CA75994-444 75994-454 75994-470
Accessory Parts for TRIO.BAS™ Air Samplers, Hardy Diagnostics
Samplers Air Samplers
An assortment of accessory and replacement products designed for use with the Hardy Diagnostics collection of TRIO.BAS™ air samplers.
Aspirating heads are available in stainless steel, Daily Shift polystyrene, or polycarbonate formats. The durable stainless steel and polycarbonate models are autoclavable and can be sprayed with disinfectant. The Daily Shift polystyrene models are supplied ready to use in a cleanroom environment. These heads are individually packaged with a Certificate of Sterility provided. The Blind Head is designed to protect the aspirating chamber when instrument is not in use.
Ordering information: Stainless steel aspirating head models BAS330 and BAS331 and induction battery charger model BAS318 are required when instruments are purchased a la carte instead of the kit format. Vertical hook and wall holder require knob (sold separately).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty