Would you like to log in again?
Synergy Nonwoven Composite Wipe, High-Tech Conversions
Supplier: High-Tech Conversions
SN-88
76169-754BG 24.34 USD InStock
76169-754
Synergy Nonwoven Composite Wipe, High-Tech Conversions
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Synergy is a tough, abrasion resistant, very absorbent, and virtually lint-free cleanroom wipe.
Synergy is an especially effective wipe. It is comprised of a synergistic blend of virgin polypropylene and cellulose that is layered and ultrasonically bonded into a uniform, super-tough layered fabric. Unique features give Synergy wipes a high degree of absorbency and effectiveness during the process of general wiping.
Packaging: Wipes are packaged 100 wipes/bag, 12 bags/case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty