Would you like to log in again?
Bee-Safe® Non-Skid Boot Cover, High-Tech Conversions
Supplier: High-Tech Conversions
GAH-BC-50
76169-756CS 481.31 USD InStock
76169-756
Bee-Safe® Non-Skid Boot Cover, High-Tech Conversions
Disposable Garments
These PE-coated, non-woven, polypropylene non-skid boot covers have an elastic leg cuff and PVC sole to protect from slips and falls.
  • Low-linting
  • Latex-free
These boot covers have a polypropylene inner layer and a polyethylene-coated outer layer. They also feature a leg cuff for comfort and stability. The PVC sole offers resistance to slips and falls against wet and smooth surfaces.
Packaging: Packaged 100 pieces/case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty