Would you like to log in again?
Wireless Nylon Racks for Microtubes, Globe Scientific
Supplier: Globe Scientific
456340Y 456340W 456345R 456345W 456345B 456340B 456340R 456345Y
76169-956EA 14.86 USD InStock
76169-956 76169-968 76169-966 76169-958 76169-970 76169-960 76169-964 76169-962
Wireless Nylon Racks for Microtubes, Globe Scientific
Racks Tube Racks
Reinforced nylon rack is autoclavable and chemical resistant.
  • Fits 1.5 and 2.0 mL microtubes
  • Single-piece construction
  • Molded handles facilitate secure handling
  • Alphanumeric markings for sample identification
  • Stackable
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty